missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Malonic acid disodium salt, 99%

Manufacturer:  Alfa Aesar™ 4484122

 View more versions of this product

Catalog No. AA4484122



Specifications

Specifications

Malonic acid disodium salt
141-95-7
MFCD00002708
PRWXGRGLHYDWPS-UHFFFAOYSA-L
disodium;propanedioate
8865
148.03
99%
100g
C3H2Na2O4
sodium malonate, disodium malonate, propanedioic acid, disodium salt, malonic acid disodium salt, disodium propanedioate, malonic acid, sodium salt, unii-9qwe64m39h, propanedioic acid, sodium salt 1:2, malonic acid, disodium salt, dicarboxymethane disodium salt
C(C(=O)[O-])C(=O)[O-].[Na+].[Na+]
148.025
CHEBI:62983
Powder

Chemical Identifiers

141-95-7
148.025
PRWXGRGLHYDWPS-UHFFFAOYSA-L
8865
disodium;propanedioate
C3H2Na2O4
MFCD00002708
sodium malonate, disodium malonate, propanedioic acid, disodium salt, malonic acid disodium salt, disodium propanedioate, malonic acid, sodium salt, unii-9qwe64m39h, propanedioic acid, sodium salt 1:2, malonic acid, disodium salt, dicarboxymethane disodium salt
CHEBI:62983
C(C(=O)[O-])C(=O)[O-].[Na+].[Na+]
Safety and Handling

Safety and Handling

RTECSNumber : OO1750000

TSCA : TSCA

SDS Documents