missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Malonic acid, 99%

Manufacturer:  Alfa Aesar™ A1152622

 View more versions of this product

Catalog No. AAA1152622



Description

Description

Applications
Malonic acid is acts as a building block in organic synthesis. It is also useful as a precursor for polyesters and alkyd resins, which is used in coating applications, thereby protecting against UV light, corrosion and oxidation. It acts as a cross linker in the coating industry and surgical adhesive. It finds application in the production of specialty chemicals, flavors and fragrances, polymer cross linkers and pharmaceuticals.

Solubility
Soluble in cold water.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
Specifications

Specifications

Malonic acid
132°C to 136°C (decomposition)
141-82-2
C3H4O4
1751370
Propanedioic acid
C(C(=O)O)C(=O)O
104.061
CHEBI:30794
157°C (314°F)
100g
99%
MFCD00002707
14,5710
OFOBLEOULBTSOW-UHFFFAOYSA-N
propanedioic acid
867
104.06

Chemical Identifiers

141-82-2
104.061
OFOBLEOULBTSOW-UHFFFAOYSA-N
867
propanedioic acid
C3H4O4
MFCD00002707
Propanedioic acid
CHEBI:30794
C(C(=O)O)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : OO0175000

TSCA : TSCA

SDS Documents