missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Malonaldehyde bis(diethyl acetal), 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 156735000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Malonaldehyde bis(diethyl acetal) | |
0.9100g/mL | |
88°C | |
97% | |
C11H24O4 | |
(C2H5O)2CHCH2CH(OC2H5)2 | |
500mL | |
02,257 | |
malonaldehyde bis diethyl acetal, tetraethoxypropane, malonaldehyde tetraethyl acetal, malonaldehyde diethyl acetal, usaf kf-26, propane, 1,1,3,3-tetraethoxy, tetraethyl malondialdehyde acetal, tetraethoxy propane, malonaldehyde tetraethyl diacetal, malonaldehyde, bis diethyl acetal | |
KVJHGPAAOUGYJX-UHFFFAOYSA-N | |
1,1,3,3-tetraethoxypropane | |
67147 | |
97% |
122-31-6 | |
220.0°C | |
Authentic | |
Glass bottle | |
1.4100 to 1.4120 | |
MFCD00009240 | |
01, III, 3095 | |
0.91 | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol,ether | |
CCOC(CC(OCC)OCC)OCC | |
220.31 | |
220.31 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 204-533-1
RUO â Research Use Only