Please login to your online account to display your discounted pricing
Thermo Scientific™ Maleimide-Activated Blue Carrier™ Protein
Make hapten-carrier immunogens comprising cysteine-containing peptides and other thiolated antigens with sulfhydryl-reactive immunogenic hemocyanin (CCH).
$128.25 - $1,092.73
Specifications
Activation Maleimide
Format Lyophilized
Protein CCH, Concholepas concholepas hemocyanin
View More Specs
Products
Catalog Number Mfr. No. Description Quantity Price Quantity    
PI77661
thermo scientific™
77661
Imject Maleimide-Activated Blue Carrier Protein 2mg Each for $128.25
PI77662
thermo scientific™
77662
Imject Maleimide-Activated Blue Carrier Protein 10 x 10mg Each for $1,092.73
PI77664
thermo scientific™
77664
Imject Maleimide-Activated Blue Carrier Protein 10mg Each for $392.86
Description & Specifications
Specifications
Activation Maleimide
Format Lyophilized
Protein CCH, Concholepas concholepas hemocyanin
Thermo Scientific™ Maleimide-Activated Blue Carrier Protein is sulfhydryl-reactive immunogenic hemocyanin (CCH) for hapten-carrier conjugation with cysteine-containing peptides and other thiolated antigens.
Blue Carrier Protein is a large homocyanin that is highly immunogenic like keyhole limpet hemocyanin (KLH) yet is more soluble. The carrier is rendered sulfhydryl-reactive by chemical modification with Sulfo-SMCC crosslinker, and the resulting maleimide-activated protein is then stabilized and lyophilized in phosphate buffer. Maleimide groups form covalent crosslinks with sulfhydryl (-SH) moieties on cysteine residues of peptides and other thiol-containing haptens. Numerous antigen molecules can be conjugated to each Blue Carrier Protein molecule to make them highly immunogenic and increase the likelihood of eliciting immunized animals to produce high-titer antisera to the epitopes of interest.
Highlights:
  • Optimized activation – specifications require that each lot is activated with more than 400 moles of sulhydryl-reactive maleimide per mole of carrier, ensuring consistently high levels of conjugatio
  • Sulfhydryl-reactive – engineered for conjugating cysteine-containing peptide antigens to make effective immunogens for high-titer antibody production
  • Simple, rapid procedure – complete the hapten-carrier conjugation reaction in two hours to yield sufficient immunogen for several injections and booster immunizations of host animals
  • High-quality carrier protein – Blue Carrier Protein is ultrapure Concholepas concholepas hemocyanin (CCH) prepared from molluscan hemolymph
  • Highly immunogenic and soluble – CCH has a high molecular mass that elicits a stronger immune response than BSA or ovalbumin