Learn More
Maleimide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 156710050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Maleimide | |
90.0°C to 94.0°C | |
Authentic | |
Glass bottle | |
MFCD00005494 | |
21,399 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol,ethylether | |
[K+].[Ag+].O=C1[N-]C(=O)C=C1.O=C1[N-]C(=O)C=C1 | |
339.10 | |
CHEBI:16072 | |
98% |
541-59-3 | |
White to Yellow | |
97.5% min. (GC) | |
C8H4AgKN2O4 | |
5 g | |
maleimide, 1h-pyrrole-2,5-dione, 2,5-pyrroledione, maleinimide, maleic imide, 3-pyrroline-2,5-dione, 2,5-dihydro-1h-pyrrole-2,5-dione, unii-2519r1ugp8, ccris 3408, 5-hydroxy-2h-pyrrol-2-one | |
KWLWLQRUFFGWNG-UHFFFAOYSA-L | |
pyrrole-2,5-dione | |
10935 | |
97.07 | |
Fine Crystalline Powder |
Chemical Identifiers
541-59-3 | |
339.10 | |
KWLWLQRUFFGWNG-UHFFFAOYSA-L | |
10935 | |
[K+].[Ag+].O=C1[N-]C(=O)C=C1.O=C1[N-]C(=O)C=C1 |
C8H4AgKN2O4 | |
MFCD00005494 | |
maleimide, 1h-pyrrole-2,5-dione, 2,5-pyrroledione, maleinimide, maleic imide, 3-pyrroline-2,5-dione, 2,5-dihydro-1h-pyrrole-2,5-dione, unii-2519r1ugp8, ccris 3408, 5-hydroxy-2h-pyrrol-2-one | |
CHEBI:16072 |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Toxic if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 208-787-4
RUO – Research Use Only