missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Maleimide, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC156715000

 View more versions of this product

Catalog No. AC156715000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Maleimide
Glass Bottle
90.0°C to 94.0°C
541-59-3
C4H3NO2
2923
1h-pyrrole-2,5-dione
PEEHTFAAVSWFBL-UHFFFAOYSA-N
pyrrole-2,5-dione
10935
97.07
98%
Authentic
White to Yellow
500g
97.5% min. (GC)
MFCD00005494
21,399
Solubility in water: soluble. Other solubilities: soluble in ethanol,ethylether
C1=CC(=O)NC1=O
97.07
CHEBI:16072
Fine Crystalline Powder

Chemical Identifiers

541-59-3
97.07
PEEHTFAAVSWFBL-UHFFFAOYSA-N
10935
pyrrole-2,5-dione
C4H3NO2
MFCD00005494
1h-pyrrole-2,5-dione
CHEBI:16072
C1=CC(=O)NC1=O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Toxic if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 208-787-4

SDS Documents