Please login to your online account to display your discounted pricing

Maleic Hydrazide, Single-Component Organic Standards, SPEX CertiPrep™

$219.83 - $219.83

Chemical Identifiers

Chemical Name or Material Maleic anhydride
CAS 108-31-6
CAS Min % 0.1
CAS Max % 0.1
CAS 67-56-1
CAS Min % 99.9
CAS Max % 99.9
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

S2357

 
spex certiprep llc
S-2357
2mL amber prescored glass ampule 1mL Each for $219.83
Description & Specifications

Specifications

Quantity 1mL
Chemical Name or Material Maleic anhydride
DOT Information Nonhazardous
Packaging 2mL amber prescored glass ampule
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Methods For US EPA Method 8270
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Recommended Storage Ambient
Solvent or Matrix Methanol-P&T
CAS 108-31-6
CAS 67-56-1
CAS Max % 99.9
CAS Max % 0.1
CAS Min % 99.9
CAS Min % 0.1

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Ambient