Please login to your online account to display your discounted pricing

Alfa Aesar™ Maleic hydrazide, 97%

$195.49 - $195.49

Chemical Identifiers

Chemical Name or Material Maleic hydrazide
CAS 123-33-1
Molecular Formula C4H4N2O2
Formula Weight 112.09
MDL Number MFCD00006665
Beilstein 112223
Merck Index 14,5705
Synonym cis-Butenedioic acid hydrazide; 1,2-Dihydro-3,6-pyridazinedione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1253136

 
alfa aesar™
A1253136
500g Each for $195.49
Description & Specifications

Specifications

Formula Weight 112.09
Molecular Formula C4H4N2O2
Chemical Name or Material Maleic hydrazide
TSCA TSCA
Assay 97%
MDL Number MFCD00006665
CAS 123-33-1
Beilstein 112223
Merck Index 14,5705
Synonym cis-Butenedioic acid hydrazide; 1,2-Dihydro-3,6-pyridazinedione
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number UR5950000
Flash Point 300°C (572°F)
Melting Point ≈305°C (dec.)
Quantity 500g

Applications
Maleic hydrazide is a synthetic plant growth regulator that is widely used for controlling growth and to induce dormancy in some crops.

Solubility
Soluble in water (4.5 g/L).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.