missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Maleate, 0.2M buffer soln., pH 6.5

$76.65 - $119.82

Chemical Identifiers

CAS 676-46-0
Molecular Formula C4H4Na2O5
Molecular Weight (g/mol) 178.051
MDL Number MFCD00012466
InChI Key WPUMTJGUQUYPIV-UHFFFAOYSA-L
PubChem CID 8736
IUPAC Name disodium;2-hydroxybutanedioate
SMILES C(C(C(=O)[O-])O)C(=O)[O-].[Na+].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61156AK View Documents Alfa Aesar™
J61156AK
250mL Each for $76.65
 
AAJ61156AP View Documents Alfa Aesar™
J61156AP
500mL Each for $119.82
 
Specifications

Chemical Identifiers

676-46-0
178.051
WPUMTJGUQUYPIV-UHFFFAOYSA-L
disodium;2-hydroxybutanedioate
C4H4Na2O5
MFCD00012466
8736
C(C(C(=O)[O-])O)C(=O)[O-].[Na+].[Na+]

Specifications

676-46-0
MFCD00012466
WPUMTJGUQUYPIV-UHFFFAOYSA-L
disodium;2-hydroxybutanedioate
8736
6.5
Maleate buffer soln.
C4H4Na2O5
0.2M buffer soln.
C(C(C(=O)[O-])O)C(=O)[O-].[Na+].[Na+]
178.051
Liquid
250mL