missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Maleate, 0.2M buffer soln., pH 6.5, Thermo Scientific™
Supplier: Thermo Scientific Chemicals J61156AP
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Maleate buffer soln. | |
0.2 M | |
Liquid | |
C4H4Na2O5 | |
WPUMTJGUQUYPIV-UHFFFAOYSA-L | |
disodium;2-hydroxybutanedioate | |
8736 |
676-46-0 | |
6.5 | |
500 mL | |
MFCD00012466 | |
C(C(C(=O)[O-])O)C(=O)[O-].[Na+].[Na+] | |
178.051 |
Chemical Identifiers
676-46-0 | |
178.051 | |
WPUMTJGUQUYPIV-UHFFFAOYSA-L | |
disodium;2-hydroxybutanedioate |
C4H4Na2O5 | |
MFCD00012466 | |
8736 | |
C(C(C(=O)[O-])O)C(=O)[O-].[Na+].[Na+] |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures