missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Maleate, 0.2M buffer soln., pH 5.0

$54.59 - $85.27

Chemical Identifiers

CAS 3105-55-3
Molecular Formula C4H9NaO7
Molecular Weight (g/mol) 192.099
MDL Number MFCD00012466
InChI Key VOWLSUZYJXDGNJ-SQDRRJMKSA-M
PubChem CID 91658932
IUPAC Name sodium;(Z)-4-hydroxy-4-oxobut-2-enoate;trihydrate
SMILES C(=CC(=O)[O-])C(=O)O.O.O.O.[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60582AK View Documents Alfa Aesar™
J60582AK
250mL Each for $54.59
 
AAJ60582AP View Documents Alfa Aesar™
J60582AP
500mL Each for $85.27
 
Specifications

Chemical Identifiers

3105-55-3
192.099
VOWLSUZYJXDGNJ-SQDRRJMKSA-M
sodium;(Z)-4-hydroxy-4-oxobut-2-enoate;trihydrate
C4H9NaO7
MFCD00012466
91658932
C(=CC(=O)[O-])C(=O)O.O.O.O.[Na+]

Specifications

3105-55-3
MFCD00012466
VOWLSUZYJXDGNJ-SQDRRJMKSA-M
sodium;(Z)-4-hydroxy-4-oxobut-2-enoate;trihydrate
91658932
5.0
250mL
C4H9NaO7
0.2M buffer soln.
C(=CC(=O)[O-])C(=O)O.O.O.O.[Na+]
192.099
Liquid
Colorless
Maleate buffer soln.