Justrite™ Maintenance Spill Berms
To contain minor spills and leaks during temporary storage of drums, machine parts, batteries and equipment
Manufacturer: Justrite™ 28424
Description
- Made of broad UV and chemical resistant PVC coated fabric
- No assembly required
- Berm anchored in place using brass grommets in each corner
- Encased urethane foam sidewalls for easy roll-through access without need for ramp
- Walls spring back into place without memory loss
- Anchor stakes not included
- Optional grates to keep products and machinery above fluid can be ordered separately
Specifications
135 gal. | |
511L (135 gal.) | |
Maintenance Spill Berm |
511L | |
3.3L x 3.04W x 0.05m H |