Please login to your online account to display your discounted pricing

GVS Maine Nylon Graph Transfer Membranes


Manufacturer: gvs maine  MM1213515

 View more versions of this product

Catalog No. NJ0HY320F5

  • / Pack of 5 Each
 

Description & Specifications

Specifications

Quantity 5/Pk.
Format Sheet
Binding Capacity 400μg/cm2
Length (Metric) 20cm
Pore Size 0.45μm
Material Nylon Graph
For Use With (Application) Radioactive and non-radioactive detection systems
Width (Metric) 20cm