Please login to your online account to display your discounted pricing

GVS Maine Nylon Charged Transfer Membranes


Manufacturer: gvs maine  MM1213449

 View more versions of this product

Catalog No. NB0HY322F5

  • / Pack of 5 Each
 

Description & Specifications

Specifications

Material Charged Nylon
Binding Capacity 450μg/cm2
Format Sheet
Pore Size 0.45μm
Length (Metric) 22cm
Width (Metric) 22cm
For Use With (Application) Southern, Northern, and alkaline transfers and reprobing procedures
Quantity 5/Pk.