Invitrogen™ eBioscience™ MagniSort™ Streptavidin Negative Selection Beads
Designed for the magnetic separation of cells by negative selection. Undesired cells are bound by a cocktail of biotinylated antibodies and then streptavidin coated magnetic beads.
Manufacturer:  Invitrogen™ LSMSNB600274
 View more versions of this product
Catalog No. 501125937
Alert:
Contains Sodium azide
Description
Description
  • Clone and concentration of biotinylated antibody for undesired cell labeling, as well as concentration of MagniSort™ Streptavidin Negative Selection Beads must be determined empirically
  • Please refer to the protocol for details on how to optimize both the antibody or antibody cocktail and MagniSort™ Streptavidin Negative Selection Beads.
  • For depletion of cells using a single antibody, we recoomend the MagniSort™ Streptavidin Positive Selection Beads (cat. MSPB-6003) and protocol
Specifications
Specifications
Magnetic Cell Separation
200 tests
2° to 8°C. Do not freeze.
Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Documents