Please login to your online account to display your discounted pricing
American Educational Products Magnetic Sphere Kit
Create visible magnetic field lines.
Manufacturer: american educational products  HUB3200
 View more versions of this product
Catalog No. S53014A
  • / Each
Description & Specifications
Specifications
For Use With (Application) Creating mini magnetic field, learning about earth magnetic core, locate magnetic pole and observe effects of magnetic field
Grade 6 to 12
Includes Sphere with an embedded Alnico magnet, iron filings, teacher's guide and petri dish
Quantity 15 Kit
Product Type Magnetic Sphere Kit