Please login to your online account to display your discounted pricing

Magnesium turnings, 99.8% (metals basis), Alfa Aesar

Form: 3.2mm (0.13 in.) wide

$54.81 - $91.91

Chemical Identifiers

Chemical Name or Material Magnesium
CAS 7439-95-4
Physical Form Turnings
Assay Percent Range 99.8% (Metals basis)
Molecular Formula Mg
Formula Weight 24.3
MDL Number MFCD00085308
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1023222

  SDS
alfa aesar
1023222
100g Each for $54.81

AA1023236

  SDS
alfa aesar
1023236
500g Each for $91.91

AA10232A4

  SDS
alfa aesar
10232A4
2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 24.3
Quantity 100g
Color Silver
DOT Information DOT Class: 4.1, Packaging Group: III
Density 1.738g/cm3 at 20°C
Physical Form Turnings
EINECS Number 231-104-6
Odor Odorless
Appearance Silver-colored turnings
Chemical Name or Material Magnesium
Assay Percent Range 99.8% (Metals basis)
Molecular Formula Mg
MDL Number MFCD00085308
CAS 7439-95-4
Health Hazard 1 H228-H251-H261: Flammable solid. Self-heating: may catch fire. In contact with water releases flammable gas.
Vapor Pressure 1.33 hPa (1mm Hg) at 621°C
Safety and Handling

H228-H251-H261: Flammable solid. Self-heating: may catch fire. In contact with water releases flammable gas.