missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Magnesium turnings, 99.8% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01023536
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Magnesium | |
Silver-colored turnings | |
99.8% (Metals basis) | |
MFCD00085308 | |
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs | |
[Mg] | |
24.30 | |
CHEBI:25107 | |
Odorless |
7439-95-4 | |
1.738g/cm3 at 20°C | |
Mg | |
500 g | |
JLVVSXFLKOJNIY-UHFFFAOYSA-N | |
magnesium | |
5462224 | |
24.3 | |
1.33 hPa (1mm Hg) at 621°C |
Chemical Identifiers
7439-95-4 | |
24.30 | |
JLVVSXFLKOJNIY-UHFFFAOYSA-N | |
5462224 | |
magnesium |
Mg | |
MFCD00085308 | |
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs | |
CHEBI:25107 | |
[Mg] |
Safety and Handling
P210-P223-P231+P232-P235+P410-P240-P241-P280-P335+P334-P370+P378s-P501c
H228-H252-H261
DOTInformation : Hazard Class: 4.1; Packaging Group: III
EINECSNumber : 231-104-6
RTECSNumber : OM2100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only