missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium turnings, 1cm (0.4 in.) & down, Puratronic™, 99.98% (metals basis)

Manufacturer:  Alfa Aesar™ 1063036

 View more versions of this product

Catalog No. AA1063036



Specifications

Specifications

Magnesium turnings
7439-95-4
Mg
UN1869
FYYHWMGAXLPEAU-UHFFFAOYSA-N
magnesium
24.305
CHEBI:25107
99.98%
500g
(metals basis)
MFCD00085308
hsdb 654
[Mg]
Odorless
5462224
1cm (0.4 in.) & down
Puratronic™

Chemical Identifiers

7439-95-4
24.305
FYYHWMGAXLPEAU-UHFFFAOYSA-N
5462224
magnesium
Mg
MFCD00085308
hsdb 654
CHEBI:25107
[Mg]
Safety and Handling

Safety and Handling

GHS H Statement
H228-H251-H261
Flammable solid.
Self-heating: may catch fire.
In contact with water releases flammable gas.

GHS P Statement
P210-P231+P232-P280-P370+P378i-P420-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store away from other materials.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: MAGNESIUM

TSCA : TSCA

Documents