Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium tungsten oxide, 99% (metals basis)

Chemical Identifiers

Chemical Name or Material Magnesium tungsten oxide
CAS 13573-11-0
Formula Weight 272.16
MDL Number MFCD00016209
Synonym Magnesium tungstate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1402314

  SDS
alfa aesar™
1402314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1402322

  SDS
alfa aesar™
1402322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 272.16
Chemical Name or Material Magnesium tungsten oxide
TSCA TSCA
Mesh Size −325 Mesh
Physical Form Powder
Assay 99%
Assay Percent Notes (metals basis)
MDL Number MFCD00016209
CAS 13573-11-0
Synonym Magnesium tungstate
Quantity 25g