missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium tungsten oxide, 99% (metals basis)

Manufacturer:  Alfa Aesar™ 1402322

 View more versions of this product

Catalog No. AA1402322



Specifications

Specifications

Magnesium tungsten oxide
(metals basis)
MFCD00016209
DJZHPOJZOWHJPP-UHFFFAOYSA-N
magnesium;dioxido(dioxo)tungsten
3084161
Powder
−325 Mesh
13573-11-0
MgO4W
magnesium tungstate, magnesium tungstate vi, magnesium tungsten oxide mgwo4, magnesium dioxido dioxo tungsten, magnesiumwolframat, magnesium wolframate, of a kind used as a luminophore, mg.wo4, tungstate wo42-, magnesium 1:1 , t-4, magnesium 2+ tungstate, magnesium tungstate-w
[O-][W](=O)(=O)[O-].[Mg+2]
272.141
272.16
99%
100g

Chemical Identifiers

13573-11-0
272.141
DJZHPOJZOWHJPP-UHFFFAOYSA-N
3084161
[O-][W](=O)(=O)[O-].[Mg+2]
MgO4W
MFCD00016209
magnesium tungstate, magnesium tungstate vi, magnesium tungsten oxide mgwo4, magnesium dioxido dioxo tungsten, magnesiumwolframat, magnesium wolframate, of a kind used as a luminophore, mg.wo4, tungstate wo42-, magnesium 1:1 , t-4, magnesium 2+ tungstate, magnesium tungstate-w
magnesium;dioxido(dioxo)tungsten
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents