missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium Trifluoromethanesulfonate, TCI America™

Manufacturer:  TCI America T130425G

 View more versions of this product

Catalog No. T130425G



Specifications

Specifications

Magnesium Trifluoromethanesulfonate
25g
C2F6MgO6S2
1759
BZQRBEVTLZHKEA-UHFFFAOYSA-L
magnesium;trifluoromethanesulfonate
2734125
Crystal-Powder at 20°C
White
60871-83-2
MFCD00042601
magnesium trifluoromethanesulfonate, magnesium triflate, magnesium 2+ ditriflate, trifluoromethanesulfonic acid magnesium salt, magnesium trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, magnesium salt, magnesiumtrifluoromethanesulfonate, magnesium trifiate, acmc-209mmv, ksc352a4p
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Mg+2]
322.431
322.43
≥98.0% (T)

Chemical Identifiers

60871-83-2
322.431
BZQRBEVTLZHKEA-UHFFFAOYSA-L
2734125
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Mg+2]
C2F6MgO6S2
MFCD00042601
magnesium trifluoromethanesulfonate, magnesium triflate, magnesium 2+ ditriflate, trifluoromethanesulfonic acid magnesium salt, magnesium trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, magnesium salt, magnesiumtrifluoromethanesulfonate, magnesium trifiate, acmc-209mmv, ksc352a4p
magnesium;trifluoromethanesulfonate
Safety and Handling

Safety and Handling

TSCA : No

Documents