Please login to your online account to display your discounted pricing

Magnesium trifluoromethanesulfonate, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  352660050

 View more versions of this product

Catalog No. AC352660050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 322.45
Quantity 5g
Color Off-white to gray
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
Infrared Spectrum Authentic
Synonym Magnesium triflate
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C2 F6 Mg O6 S2
Chemical Name or Material Magnesium trifluoromethanesulfonate
MDL Number MFCD00042601
CAS 60871-83-2

Specifications

Chemical Name or Material Magnesium trifluoromethanesulfonate
Name Note 98%
CAS 60871-83-2
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C2 F6 Mg O6 S2
Formula Weight 322.45
MDL Number MFCD00042601
Synonym Magnesium triflate
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents