Please login to your online account to display your discounted pricing

Magnesium tert-butoxide, +93%, ACROS Organics™

$144.17 - $144.17

Chemical Identifiers

Chemical Name or Material Magnesium tert-butoxide
CAS 32149-57-8
Assay ≥93.0%
Assay Percent Range 93+%
Molecular Formula C8H18MgO2
Formula Weight 170.53
MDL Number MFCD03788761
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC390470100

  SDS
thermo fisher scientific
AC390470100
Glass Bottle 10g Each for $144.17

AC390470500

  SDS
thermo fisher scientific
AC390470500
Plastic bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC390472500

  SDS
thermo fisher scientific
AC390472500
Plastic bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥93.0%
Physical Form Powder
Formula Weight 170.53
Quantity 10g
Color White to light gray
Packaging Glass Bottle
Chemical Name or Material Magnesium tert-butoxide
Assay Percent Range 93+%
MDL Number MFCD03788761
CAS 32149-57-8
Health Hazard 2 GHS H Statement:
Flammable solid.
Causes severe skin burns and eye damage.
Reacts violently with water.
Molecular Formula C8H18MgO2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable solid.
Causes severe skin burns and eye damage.
Reacts violently with water.