missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium tert-butoxide, 90+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1974518

 View more versions of this product

Catalog No. AAL1974518



Description

Description

Magnesium tert-butoxide is used as a pharmaceutical intermediate. It is also used in deprotonation reactions for organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium tert-butoxide is used as a pharmaceutical intermediate. It is also used in deprotonation reactions for organic syntheses.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with bases and oxidizing agents. Store under dry inert gas. It is sensitive to air and moisture.
Specifications

Specifications

Magnesium tert-butoxide
C8H18MgO2
MFCD03788761
UN2921
Air and moisture sensitive
Reacts with water.
CC(C)(C)[O-].CC(C)(C)[O-].[Mg+2]
170.535
170.54
32149-57-8
[(CH3)3CO]2Mg
50 g
3979348
magnesium tert-butoxide, magnesium di-tert-butoxide, magnesium 2-methylpropan-2-olate, magnesium t-butoxide, magnesiumtert-butoxide, di-tert-butoxymagnesium, magnesium tert. butoxide, ksc226k7n, magnesium 2+ bis 2-methylpropan-2-olate
RTKCPZYOLXPARI-UHFFFAOYSA-N
magnesium;2-methylpropan-2-olate
12040442
≥90%

Chemical Identifiers

32149-57-8
170.535
RTKCPZYOLXPARI-UHFFFAOYSA-N
12040442
CC(C)(C)[O-].CC(C)(C)[O-].[Mg+2]
C8H18MgO2
MFCD03788761
magnesium tert-butoxide, magnesium di-tert-butoxide, magnesium 2-methylpropan-2-olate, magnesium t-butoxide, magnesiumtert-butoxide, di-tert-butoxymagnesium, magnesium tert. butoxide, ksc226k7n, magnesium 2+ bis 2-methylpropan-2-olate
magnesium;2-methylpropan-2-olate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H228
Causes severe skin burns and eye damage.
Flammable solid.

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H319-H332-H335

DOTInformation : Hazard Class: 4.2; Packaging Group: II

EINECSNumber : 250-931-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only