Please login to your online account to display your discounted pricing

Magnesium Sulfate USP/FCC, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Magnesium Sulfate
Name Note 7-Hydrate
CAS 10034-99-8
Molecular Formula MgSO4·7H2O
Formula Weight 246.47
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK4200P212

 
avantor performance
420007
Poly 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade FCC/USP
Formula Weight 246.47
Quantity 2.5kg
Chemical Name or Material Magnesium Sulfate
CAS 10034-99-8
Molecular Formula MgSO4·7H2O
Packaging Poly
Name Note 7-Hydrate