missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium sulfate hydrate, Puratronic™, 99.997% (metals basis)

Manufacturer:  Alfa Aesar™ 1080118

 View more versions of this product

Catalog No. AA1080118



Specifications

Specifications

Magnesium sulfate hydrate
(metals basis)
MFCD00149787
bitter salt
[O-]S(=O)(=O)[O-].[Mg+2]
120.361
CHEBI:32599
Crystalline Aggregates
Puratronic™
Odorless
50g
22189-08-8
MgO4S
14,5691
CSNNHWWHGAXBCP-UHFFFAOYSA-L
magnesium;sulfate
24083
120.37 (Anhydrous)
99.997%
1.68g/mL
White

Chemical Identifiers

22189-08-8
120.361
CSNNHWWHGAXBCP-UHFFFAOYSA-L
24083
magnesium;sulfate
MgO4S
MFCD00149787
bitter salt
CHEBI:32599
[O-]S(=O)(=O)[O-].[Mg+2]
Safety and Handling

Safety and Handling

RTECSNumber : OM4500000

TSCA : TSCA

Documents