Please login to your online account to display your discounted pricing

Magnesium Sulfate Heptahydrate (USP/FCC), Fisher Chemical


Quantity 50kg
Packaging Fiber Drum

Manufacturer: fisher scientific company  M6750

 View more versions of this product

Catalog No. M67-50

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade FCC/USP
Formula Weight 246.48
Melting Point 770°C
Vapor Pressure <0.1 mmHg at 20°C
Quantity 50kg
Assay 99.0 to 100.5% (USP)
Physical Form Solid
Molecular Formula MgO4S · 7 H2O
Color White
pH 5 to 8.2
Density 1.678g/cm³
Chemical Name or Material Magnesium Sulfate Heptahydrate
Identification Pass Test (FCC)
Iron (Fe) 20ppm max. (USP)
Lead (Pb) 4mg/kg max. (FCC)
Loss on Ignition 40.0 to 52.0% (FCC)
Packaging Fiber Drum
Selenium (Se) 0.003% max. (FCC)
MDL Number MFCD00149785
CAS 10034-99-8
ChemAlert Storage Symbol Gray
Heavy Metals (as Pb) 0.001% max. (USP)
Residual Solvents Meets Requirements (USP)
Synonym Epsom Salts
Chloride 0.014% max. (USP)

Specifications

Chemical Name or Material Magnesium Sulfate Heptahydrate
CAS 10034-99-8
Purity Grade FCC/USP
Assay 99.0 to 100.5% (USP)
Molecular Formula MgO4S · 7 H2O
Formula Weight 246.48
MDL Number MFCD00149785
Synonym Epsom Salts
Accessories
 

SDS & Safety Documents