Please login to your online account to display your discounted pricing

Magnesium Sulfate Heptahydrate (USP/FCC), Fisher Chemical

$917.36 - $2,973.21

Chemical Identifiers

Chemical Name or Material Magnesium Sulfate Heptahydrate
CAS 10034-99-8
Purity Grade FCC/USP
Assay 99.0 to 100.5% (USP)
Molecular Formula MgO4S · 7 H2O
Formula Weight 246.48
MDL Number MFCD00149785
Synonym Epsom Salts
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M67-3

  SDS
fisher scientific company
M673
Glass Bottle 3kg Each for $917.36
Case of 4 Each for $3,669.44

M67-50

  SDS
fisher scientific company
M6750
Fiber Drum 50kg Each for $2,973.21
Description & Specifications

Specifications

Assay ≥99.5% (FCC)
Purity Grade FCC/USP
Formula Weight 246.48
Melting Point 770°C
Vapor Pressure <0.1 mmHg at 20°C
Quantity 3kg
Physical Form Solid
Molecular Formula MgO4S · 7 H2O
Color White
pH 5 to 8.2
Density 1.678g/cm³
Identification Pass Test (FCC)
Loss on Ignition 40.0 to 52.0% (FCC)
Packaging Glass Bottle
Selenium (Se) 0.003% max. (FCC)
MDL Number MFCD00149785
Synonym Epsom Salts
Chemical Name or Material Magnesium Sulfate Heptahydrate
Heavy Metals (as Pb) 0.001% max. (USP)
Iron (Fe) 20ppm max. (USP)
Lead (Pb) 4mg/kg max. (FCC)
Residual Solvents Meets Requirements (USP)
CAS 10034-99-8
ChemAlert Storage Symbol Gray
Chloride 0.014% max. (USP)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling