missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium sulfate heptahydrate, Cell Culture Reagent

Manufacturer:  Alfa Aesar™ J6183936

 View more versions of this product

Catalog No. AAJ6183936



Specifications

Specifications

10034-99-8
MgSO4·7H2O
14,5691
WRUGWIBCXHJTDG-UHFFFAOYSA-L
magnesium;sulfate;heptahydrate
24843
246.48 (120.37 Anhydrous)
Cell Culture Reagent
Odorless
500g
H14MgO11S
MFCD00149785
magnesium ii , sulfate, heptahydrate, magnesium sulfate 1:1 heptahydrate, magnesium sulfate heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, magnesium sulfate usan:jan, magnesium sulphate heptahydrate, mgso4.7h2o, sulfuric acid magnesium salt 1:1 , heptahydrate, sulfuric acid, magnesium salt 1:1 , heptahydrate, unii-sk47b8698t
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Mg+2]
246.466
CHEBI:31795
Powder
1.68
White

Chemical Identifiers

10034-99-8
246.466
WRUGWIBCXHJTDG-UHFFFAOYSA-L
24843
magnesium;sulfate;heptahydrate
H14MgO11S
MFCD00149785
magnesium ii , sulfate, heptahydrate, magnesium sulfate 1:1 heptahydrate, magnesium sulfate heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, magnesium sulfate usan:jan, magnesium sulphate heptahydrate, mgso4.7h2o, sulfuric acid magnesium salt 1:1 , heptahydrate, sulfuric acid, magnesium salt 1:1 , heptahydrate, unii-sk47b8698t
CHEBI:31795
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Mg+2]
Safety and Handling

Safety and Handling

EINECSNumber : 231-298-2

RTECSNumber : OM4508000

TSCA : No

Recommended Storage : Ambient temperatures

Documents