Please login to your online account to display your discounted pricing

Magnesium Sulfate, Heptahydrate, ACS Grade, 98.0 to 102.0 %, LabChem™

$153.49 - $153.49

Chemical Identifiers

Chemical Name or Material Magnesium Sulfate
CAS 10034-99-8
CAS Max % 100
Name Note Heptahydrate
Purity Grade ACS Grade
Linear Formula MgSO4·7H2O
Formula Weight 246.47
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC164901

 
lab chem
LC164901
500g Each for $153.49
Description & Specifications

Specifications

Assay 98.0 to 102.0%
Formula Weight 246.47
Chemical Name or Material Magnesium Sulfate
Identification Passes Test
Name Note Heptahydrate
Packaging Poly Bottle
Purity Grade ACS Grade
Physical Form Crystals
Solubility Soluble in water
Storage Information Room Temperature
Linear Formula MgSO4·7H2O
CAS 10034-99-8
Decomposition Information Sulfur compounds
Color White
Quantity 500g
CAS Max % 100
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.