missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium Sulfate, Heptahydrate, 98.0 to 102.0%, ACS, EMD Millipore

Manufacturer:  Millipore Sigma MX00703

 View more versions of this product

Catalog No. MMX00703



Specifications

Specifications

Magnesium Sulfate, Heptahydrate
10034-99-8
H14MgO11S
magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Mg+2]
246.466
CHEBI:31795
Crystalline Solid
900kg/m3
White
EMSURE™, For Analysis
98.0 to 102.0%
MgSO4 · 7H2O
WRUGWIBCXHJTDG-UHFFFAOYSA-L
magnesium;sulfate;heptahydrate
24843
246.48
ACS/EP/Reagent
1.68g/cm3 (20°C)
2.5kg

Chemical Identifiers

10034-99-8
246.466
magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
CHEBI:31795
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Mg+2]
H14MgO11S
WRUGWIBCXHJTDG-UHFFFAOYSA-L
24843
magnesium;sulfate;heptahydrate
Safety and Handling

Safety and Handling

Recommended Storage : Store without restrictions

Documents