Please login to your online account to display your discounted pricing

Magnesium Sulfate, Heptahydrate, 98.0 to 102.0%, ACS, EMD Millipore

$131.43 - $476.00

Chemical Identifiers

Chemical Name or Material Magnesium Sulfate, Heptahydrate
CAS 10034-99-8
Name Note EMSURE™, For Analysis
Purity Grade ACS/EP/Reagent Grade
Molecular Formula MgO4S · 7H2O
Linear Formula MgSO4 · 7H2O
Formula Weight 246.48
Synonym Bitter salt
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

MMX00703

 
emd millipore corporation
MX00703
Each for $476.00

MMX00701

 
emd millipore corporation
MX00701
Each for $131.43
Description & Specifications

Specifications

Formula Weight 246.48
Physical Form Crystalline Solid
Purity Grade ACS/EP/Reagent Grade
Color White
Quantity 2.5kg
Bulk Density 900kg/m3
Density 1.68g/cm3 (20°C)
Synonym Bitter salt
Recommended Storage Store without restrictions
Chemical Name or Material Magnesium Sulfate, Heptahydrate
Assay Percent Range 98.0 to 102.0%
Name Note EMSURE™, For Analysis
CAS 10034-99-8
Molecular Formula MgO4S · 7H2O
Linear Formula MgSO4 · 7H2O
Safety and Handling

Store without restrictions