missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Magnesium sulfate, dried, contains ca 1-2 mol water of hydration, Thermo Scientific Chemicals
$46.07 - $142.11
Chemical Identifiers
CAS | 22189-08-8 |
---|---|
Molecular Formula | MgO4S |
Molecular Weight (g/mol) | 120.36 |
MDL Number | MFCD00149787 |
InChI Key | CSNNHWWHGAXBCP-UHFFFAOYSA-L |
Synonym | magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot |
PubChem CID | 24083 |
ChEBI | CHEBI:32599 |
IUPAC Name | magnesium;sulfate |
SMILES | [Mg++].[O-]S([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1373922
|
Thermo Scientific Chemicals
L1373922 |
100 g |
Each for $46.07
|
|
|||||
AAL1373936
|
Thermo Scientific Chemicals
L1373936 |
500 g |
Each for $79.00
|
|
|||||
AAL137390E
|
Thermo Scientific Chemicals
L137390E |
2500 g |
Each for $142.11
|
|
|||||
Description
Suitable for the preliminary drying of organic liquids, for example solvent extracts from reaction workups.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
22189-08-8 | |
120.36 | |
CSNNHWWHGAXBCP-UHFFFAOYSA-L | |
24083 | |
magnesium;sulfate |
MgO4S | |
MFCD00149787 | |
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot | |
CHEBI:32599 | |
[Mg++].[O-]S([O-])(=O)=O |
Specifications
22189-08-8 | |
MgO4S | |
14,5691 | |
CSNNHWWHGAXBCP-UHFFFAOYSA-L | |
magnesium;sulfate | |
24083 | |
120.37 (Anhydrous) | |
Magnesium sulfate, contains ≈1 to 2 mol water of hydration |
100 g | |
MFCD00149787 | |
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot | |
[Mg++].[O-]S([O-])(=O)=O | |
120.36 | |
CHEBI:32599 | |
Hygroscopic |
Safety and Handling
EINECSNumber : 231-298-2
RTECSNumber : OM4500000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only