missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium Sulfate AR Powder, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 7487-88-9
Physical Form Powder
Molecular Formula MgSO4
Formula Weight 120.37
Chemical Name or Material Magnesium Sulfate, Anhydrous
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK6070500 Avantor Performance
607012
POLYSTORMOR 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7487-88-9
MgSO4
Magnesium Sulfate, Anhydrous
Powder
120.37

Specifications

7487-88-9
MgSO4
POLYSTORMOR
Magnesium Sulfate, Anhydrous
Powder
120.37
500g