missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Magnesium Sulfate, 2.25% (w/v), Ricca Chemical
Supplier: Ricca Chemical Company 453016
Specifications
Magnesium Sulfate | |
10034-99-8,7732-18-5 | |
MFCD00149785 | |
WRUGWIBCXHJTDG-UHFFFAOYSA-L | |
magnesium(2+) heptahydrate sulfate | |
24843 | |
Natural Poly Bottle | |
500 mL | |
2.25% (w/v) |
Colorless | |
H14MgO11S | |
magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate | |
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O | |
246.47 | |
CHEBI:31795 | |
Aqueous Solution | |
Liquid |
Chemical Identifiers
10034-99-8 | |
246.47 | |
WRUGWIBCXHJTDG-UHFFFAOYSA-L | |
24843 | |
magnesium(2+) heptahydrate sulfate |
H14MgO11S | |
MFCD00149785 | |
magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate | |
CHEBI:31795 | |
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O |