Please login to your online account to display your discounted pricing

Magnesium Sulfate, 2.25% (w/v) Aqueous Solution, For APHA 5-Day BOD Test, Ricca Chemical


Quantity 20L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR453000020F

 View more versions of this product

Catalog No. R453000020F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Magnesium Sulfate
Name Note 2.25% w/v
CAS 7732-18-5
CAS 10034-99-8
Physical Form Liquid
Assay Percent Range 2.250 ± 0.003% (w/v)
Odor Odorless
Packaging Cubitainer
Solvent or Matrix Aqueous Solution
Color Clear colorless
Quantity 20L

Specifications

Chemical Name or Material Magnesium Sulfate
Name Note 2.25% w/v
CAS 7732-18-5
CAS 10034-99-8
Physical Form Liquid
Assay Percent Range 2.250 ± 0.003% (w/v)