Please login to your online account to display your discounted pricing

Magnesium Sulfate, 2.25% (w/v) Aqueous Solution, For APHA 5-Day BOD Test, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Magnesium Sulfate
Name Note 2.25% w/v
CAS 7732-18-5
CAS 10034-99-8
Physical Form Liquid
Assay Percent Range 2.250 ± 0.003% (w/v)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R453000020F

 
ricca chemical company
RIR453000020F
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R45300004A

 
ricca chemical company
RIR45300004A
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 20L
Solvent or Matrix Aqueous Solution
Chemical Name or Material Magnesium Sulfate
Assay Percent Range 2.250 ± 0.003% (w/v)
CAS 7732-18-5
CAS 10034-99-8
Packaging Cubitainer
Name Note 2.25% w/v
Odor Odorless