missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium sulfate, 1M aq. soln., sterile

$76.49 - $114.89

Chemical Identifiers

CAS 7487-88-9
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.361
MDL Number MFCD00011110
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
PubChem CID 24083
ChEBI CHEBI:32599
IUPAC Name magnesium;sulfate
SMILES [O-]S(=O)(=O)[O-].[Mg+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61030EQE View Documents Alfa Aesar™
J61030EQE
125mL Each for $76.49
 
AAJ61030AK View Documents Alfa Aesar™
J61030AK
250mL Each for $114.89
 
Description

Description

Solubility
Fully miscible in water. Soluble in alcohol, ether (11.6 mg/ml at 18°C) and glycerin. Insoluble in acetone.

Notes
Stable under recommended storage conditions. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

7487-88-9
120.361
CSNNHWWHGAXBCP-UHFFFAOYSA-L
24083
magnesium;sulfate
MgO4S
MFCD00011110
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
CHEBI:32599
[O-]S(=O)(=O)[O-].[Mg+2]

Specifications

7487-88-9
MgSO4
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
[O-]S(=O)(=O)[O-].[Mg+2]
120.361
CHEBI:32599
Liquid
125mL
MgO4S
MFCD00011110
CSNNHWWHGAXBCP-UHFFFAOYSA-L
magnesium;sulfate
24083
120.37
1M aq. soln.
Magnesium sulfate, sterile
Safety and Handling

Safety and Handling

TSCA : TSCA