Please login to your online account to display your discounted pricing

Magnesium Stearate (Impalpable Powder/NF), Fisher Chemical

$300.29 - $652.40

Chemical Identifiers

Chemical Name or Material Magnesium Stearate
CAS 557-04-0
Purity Grade NF
Physical Form Solid
Assay 4 to 5%
Molecular Formula C36H70MgO4
Linear Formula Mg(C18H35O2)2
Formula Weight 591.27
MDL Number MFCD00036391
Synonym Octadecanoic Acid Magnesium Salt
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M58-500

  SDS
fisher scientific company
FLM58500
Poly Bottle 500g Each for $300.29
Case of 4 Each for $1,201.16

M58-212

  SDS
thermo fisher scientific
FLM58212
Poly Pail 2.5kg Each for $652.40

M58-50

  SDS
fisher scientific company
FLM5850
Fiber Drum 50kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 591.27
Melting Point 130°C
Quantity 500g
Purity Grade NF
Physical Form Solid
Assay 4 to 5%
Molecular Formula C36H70MgO4
Color White
MDL Number MFCD00036391
Residual Solvents Meets Requirements
Stearic and Palmitic Acid 90% min.
Sulfate 1% max.
Acidity Alkalinity Pass Test
Escherichia coli (E coli) Negative Negative/Positive
Identification Pass Test
ChemAlert Storage Symbol Gray
Lead (Pb) 0.001% max.
Synonym Octadecanoic Acid Magnesium Salt
Loss on Drying 6% max.
Packaging Poly Bottle
Salmonella Negative Positive/Negative
Stearic Acid 40% min.
Total Microbial Count 1000 PER 1000 max.
Total Mold and Yeast 500 PER 500 max.
Chloride 0.1% max.
Chemical Name or Material Magnesium Stearate
Linear Formula Mg(C18H35O2)2
CAS 557-04-0

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling