Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.95% (metals basis)

$675.74 - $675.74

Chemical Identifiers

Chemical Name or Material Magnesium sputtering target
CAS 7439-95-4
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00085308
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40861KS

 
alfa aesar™
40861KS
1 each Each for $675.74
Description & Specifications

Specifications

Chemical Name or Material Magnesium sputtering target
TSCA TSCA
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
Assay 99.95%
Assay Percent Notes (metals basis)
MDL Number MFCD00085308
CAS 7439-95-4
Odor Odorless
Quantity 1 each