Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.95% (metals basis)

$382.72 - $382.72

Chemical Identifiers

Chemical Name or Material Magnesium sputtering target
CAS 7439-95-4
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00085308
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40859KS

  SDS
alfa aesar™
40859KS
1 each Each for $382.72
Description & Specifications

Specifications

Chemical Name or Material Magnesium sputtering target
Assay Percent Notes (metals basis)
Odor Odorless
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
TSCA TSCA
MDL Number MFCD00085308
Assay 99.95%
CAS 7439-95-4
Quantity 1 each