missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium Slug, Dia. X L: 3.175 x 3.175mm (0.125 x 0.125 in.), 99.95% (Metals basis), Alfa Aesar™

Form: Approx.0.04g/slug

Manufacturer:  Alfa Aesar 4329618

 View more versions of this product

Catalog No. AA4329618



Specifications

Specifications

Magnesium
Unannealed
Silver-colored slug
99.95% (Metals basis)
MFCD00085308
FYYHWMGAXLPEAU-UHFFFAOYSA-N
magnesium
24.305
CHEBI:25107
Silver
50g
7439-95-4
1.738g/cm3 at 20°C
Mg
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
[Mg]
Odorless
5462224
Slug, dia. X L: 3.175 X 3.175mm (0.125 X 0.125 in.)

Chemical Identifiers

7439-95-4
24.305
FYYHWMGAXLPEAU-UHFFFAOYSA-N
5462224
magnesium
Mg
MFCD00085308
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
CHEBI:25107
[Mg]
Documents