missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 4329830

 View more versions of this product

Catalog No. AA4329830



Specifications

Specifications

Magnesium slug
250g
7439-95-4
Mg
hsdb 654
[Mg]
Odorless
5462224
Slug
≈0.35g/slug
6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length
(metals basis)
MFCD00085308
FYYHWMGAXLPEAU-UHFFFAOYSA-N
magnesium
24.305
CHEBI:25107
99.95%

Chemical Identifiers

7439-95-4
24.305
FYYHWMGAXLPEAU-UHFFFAOYSA-N
5462224
magnesium
Mg
MFCD00085308
hsdb 654
CHEBI:25107
[Mg]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents