Please login to your online account to display your discounted pricing

Magnesium silicide, Blue 3-12mm Pieces, 99.99% (Metals basis), Alfa Aesar™

Air and Moisture Sensitive; Decomposes in water, HCl

Chemical Identifiers

Chemical Name or Material Magnesium silicide
CAS 22831-39-6
Physical Form 3-12mm Pieces
Assay Percent Range 99.99% (Metals basis)
Molecular Formula Mg2Si
Formula Weight 76.71
MDL Number MFCD00016202
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4551809

 
alfa aesar
4551809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4551818

 
alfa aesar
4551818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4551830

 
alfa aesar
4551830
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form 3-12mm Pieces
Formula Weight 76.71
Quantity 10g
Color Blue
DOT Information DOT Class: 4.3, Packaging Group: II
EINECS Number 245-254-5
Molecular Formula Mg2Si
Chemical Name or Material Magnesium silicide
Assay Percent Range 99.99% (Metals basis)
MDL Number MFCD00016202
CAS 22831-39-6
Health Hazard 1 H261: In contact with water releases flammable gas.
Safety and Handling

H261: In contact with water releases flammable gas.