missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium silicide, 99.5% (metals basis), Alfa Aesar™

Air and Moisture Sensitive; Decomposes in water, HCl

Manufacturer:  Thermo Fisher Scientific 1283718

 View more versions of this product

Catalog No. AA1283718



Specifications

Specifications

Magnesium silicide
99.5% (Metal basis)
MFCD00016202
dimagnesium monosilicide
YTHCQFKNFVSQBC-UHFFFAOYSA-N
76.695
76.71
2 g/cm3 at 20°C
Blue
50g
22831-39-6
Mg2Si
Blue powder
Insoluble in water and denser than water.
[Mg]=[Si]=[Mg]
89858
-20 Mesh Powder
Air and moisture sensitive
1102°C

Chemical Identifiers

22831-39-6
76.695
YTHCQFKNFVSQBC-UHFFFAOYSA-N
89858
Mg2Si
MFCD00016202
dimagnesium monosilicide
[Mg]=[Si]=[Mg]
Safety and Handling

Safety and Handling

P223-P231+P232-P280-P335+P334-P370+P378q-P501c

H261

DOTInformation : DOT Class: 4.3, Packaging Group: II

EINECSNumber : 245-254-5

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents