missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium silicide, 99.5% (metals basis), Alfa Aesar™

Air and Moisture Sensitive; Decomposes in water, HCl

Manufacturer:  Alfa Aesar 1283709

 View more versions of this product

Catalog No. AA1283709



Specifications

Specifications

Magnesium silicide
99.5% (Metal basis)
MFCD00016202
magnesium silicide, magnesium silicide, dimagnesium silicide, dimagnesium monosilicide, unii-475e6fmg3k, magnesiumsilicid, magnesium, compd. with silicon 2:1, magnesium silicide, powder, magnesium silicide-45 my, magnesium silicide un2624 dangerous when wet
[Mg]=[Si]=[Mg]
89858
Powder
10g
22831-39-6
Mg2Si
Blue powder
YTHCQFKNFVSQBC-UHFFFAOYSA-N
76.695
76.71
Blue

Chemical Identifiers

22831-39-6
76.695
YTHCQFKNFVSQBC-UHFFFAOYSA-N
89858
Mg2Si
MFCD00016202
magnesium silicide, magnesium silicide, dimagnesium silicide, dimagnesium monosilicide, unii-475e6fmg3k, magnesiumsilicid, magnesium, compd. with silicon 2:1, magnesium silicide, powder, magnesium silicide-45 my, magnesium silicide un2624 dangerous when wet
[Mg]=[Si]=[Mg]
Safety and Handling

Safety and Handling

H261; In contact with water releases flammable gas.

DOTInformation : DOT Class: 4.3, Packaging Group: II

EINECSNumber : 245-254-5

Documents