missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium silicate, 99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4380718

 View more versions of this product

Catalog No. AA4380718



Specifications

Specifications

Magnesium silicate
99% (Metal basis)
MFCD00078249
ZADYMNAVLSWLEQ-UHFFFAOYSA-N
140.71
3.21g/cm3 at 20°C
White
1343-88-0
MgO3Si
White powder
100.39
Powder
Odorless
50g

Chemical Identifiers

1343-88-0
100.39
ZADYMNAVLSWLEQ-UHFFFAOYSA-N
MgO3Si
MFCD00078249
Safety and Handling

Safety and Handling

H332: Harmful if inhaled.

EINECSNumber : 215-681-1

Documents