missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium rod, 7.9mm (0.31 in.) dia. x 25mm (1 in.) length, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 43355KM

 View more versions of this product

Catalog No. AA43355KM



Specifications

Specifications

Magnesium rod
25 pc.
7439-95-4
Mg
hsdb 654
[Mg]
Odorless
5462224
Rod
≈2.1g/piece
7.9mm (0.31 in.) dia. x 25mm (1 in.) length
(metals basis)
MFCD00085308
FYYHWMGAXLPEAU-UHFFFAOYSA-N
magnesium
24.305
CHEBI:25107
99.9%

Chemical Identifiers

7439-95-4
24.305
FYYHWMGAXLPEAU-UHFFFAOYSA-N
5462224
magnesium
Mg
MFCD00085308
hsdb 654
CHEBI:25107
[Mg]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents