missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Magnesium rod, 6.35mm (0.25in) dia x 25.4mm (1.0in) length, 99.95% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 047073KF
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Magnesium rod | |
7439-95-4 | |
Mg | |
1 Pc. | |
JLVVSXFLKOJNIY-UHFFFAOYSA-N | |
magnesium | |
5462224 | |
99.95% | |
≈1.4g/piece |
6.35mm (0.25 in.) dia. x 25.4mm (1.0 in.) length | |
(metals basis) | |
MFCD00085308 | |
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs | |
[Mg] | |
24.30 | |
CHEBI:25107 | |
Odorless |
Chemical Identifiers
7439-95-4 | |
24.30 | |
JLVVSXFLKOJNIY-UHFFFAOYSA-N | |
5462224 | |
magnesium |
Mg | |
MFCD00085308 | |
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs | |
CHEBI:25107 | |
[Mg] |
Safety and Handling
P210-P223-P231+P232-P235+P410-P240-P241-P280-P335+P334-P370+P378s-P501c
H228-H252-H261
DOTInformation : Hazard Class: 4.1; Packaging Group: III
EINECSNumber : 231-104-6
RTECSNumber : OM2100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only